(Trifluoromethyl)tris(triphenylphosphine)copper(I) 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (Trifluoromethyl)tris(triphenylphosphoranyl)copper, Tris(triphenylphosphine)(trifluoromethyl)copper(I)

A348007-1G A348007-250MG A348007-5G A348007-25G
77015-690EA 43 USD
77015-690 77015-692 77015-694 77684-027
(Trifluoromethyl)tris(triphenylphosphine)copper(I) 98%, Ambeed.Inc
(Trifluoromethyl)tris(triphenylphosphine)copper(I)

Precaución: For research use only.

Formula: C₅₅H₄₈CuF₃P₃
MW: 922.43 g/mol
MDL Number: MFCD22200540
CAS Number: 325810-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR