2,5-Dichlorophenethyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2,5-Dichlorophenyl)ethan-1-ol

A347966-10G A347966-1G A347966-25G A347966-5G
77015-682EA 41 USD
77015-682 77015-684 77015-686 77015-688
2,5-Dichlorophenethyl alcohol 97%
2,5-Dichlorophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O
MDL Number: MFCD04038828
CAS Number: 1875-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR