3-Bromoquinolin-7-amine 95%

Supplier: Ambeed
Warning

A347435-100MG A347435-250MG A347435-1G
76989-550EA 31 USD
76989-550 76989-552 77572-012
3-Bromoquinolin-7-amine 95%
3-Bromoquinolin-7-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD20327980
CAS Number: 1344046-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR