4-Chloro-2-methoxy-3-nitropyridine 97%
Supplier: Ambeed
|
Warning
|
A347306-100MG
A347306-250MG
A347306-1G
76989-546EA
34
USD
76989-546
76989-548
77594-036
4-Chloro-2-methoxy-3-nitropyridine 97%
4-Chloro-2-methoxy-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₃ |
MDL Number:
MFCD17014990 CAS Number: 934180-48-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...