2-Chloro-6,7-dimethoxyquinoline-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A347256-250MG A347256-5G
76989-542EA 24.41 USD
76989-542 76989-544
2-Chloro-6,7-dimethoxyquinoline-3-carbaldehyde 95%
2-Chloro-6,7-dimethoxyquinoline-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO₃
MW: 251.67 g/mol
MDL Number: MFCD02049246
CAS Number: 68236-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR