(5-(Benzyloxy)-2-formylphenyl)boronic acid 96%
Supplier: Ambeed
|
Warning
|
A347195-100MG
A347195-1G
A347195-250MG
77015-676EA
38.2
USD
77015-676
77015-678
77015-680
(5-(Benzyloxy)-2-formylphenyl)boronic acid 96%
(5-(Benzyloxy)-2-formylphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₄H₁₃BO₄ MW: 256.06 g/mol |
MDL Number:
MFCD18383732 CAS Number: 1226773-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...