(S)-2-((tert-Butoxycarbonyl)amino)-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid 95%
Supplier: Ambeed
|
Warning
|
A346896-100MG
A346896-250MG
A346896-1G
77003-208EA
128
USD
77003-208
77594-034
77601-550
(S)-2-((tert-Butoxycarbonyl)amino)-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid 95%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(1H-pyrrolo[2,3-b]pyridin-3-yl)propanoic acid
Precaución: for Research Use Only
Formula:
C₁₅H₁₉N₃O₄ |
MDL Number:
MFCD08273059 CAS Number: 146610-21-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...