cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A346802-100MG A346802-1G A346802-250MG A346802-5G
77015-668EA 19.1 USD
77015-668 77015-670 77015-672 77015-674
cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate 95%
cis-tert-Butyl 3-amino-4-fluoropyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₇FN₂O₂
MW: 204.24 g/mol
MDL Number: MFCD18072719
CAS Number: 1431720-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR