2,3-Difluoro-5-nitrobenzonitrile 97%
Supplier: Ambeed
|
Warning
|
A346412-100MG
A346412-1G
A346412-250MG
A346412-50MG
76989-530EA
81
USD
76989-530
76989-532
76989-534
76989-536
2,3-Difluoro-5-nitrobenzonitrile 97%
2,3-Difluoro-5-nitrobenzonitrile
Precaución: for Research Use Only
Formula:
C₇H₂F₂N₂O₂ |
MDL Number:
MFCD24038781 CAS Number: 1247885-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...