5-Bromo-2-(methylthio)pyrimidine 98%

Supplier: Ambeed
Danger

Synonyms: 5-Bromo-2-methylthiopyrimidine

A346162-100G A346162-10G A346162-1G A346162-250MG A346162-25G
77015-648EA 491.2 USD
77015-648 77015-650 77015-652 77015-654 77015-656
5-Bromo-2-(methylthio)pyrimidine 98%
5-Bromo-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂S
MW: 205.0756 g/mol
MDL Number: MFCD00187875
CAS Number: 14001-67-3
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR