Fedratinib (TG101348) 98%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butyl)-3-((5-methyl-2-((4-(2-(pyrrolidin-1-yl)ethoxy)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide, Fedratinib, SAR-302503

A345883-1MG A345883-5MG A345883-1G A345883-250MG A345883-100MG A345883-10MG
77033-082EA 12.74 USD
77033-082 77572-008 77601-538 77601-540 77668-582 77668-584
Fedratinib (TG101348) 98%
Fedratinib (TG101348)

Precaución: for Research Use Only

Formula: C₂₇H₃₆N₆O₃S
MW: 524.7 g/mol
MDL Number: MFCD12922515
CAS Number: 936091-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR