Pyrrol-1-ylcarbamic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 1H-pyrrol-1-ylcarbamate

A345726-100G A345726-25G A345726-5G
77033-076EA 206.88 USD
77033-076 77033-078 77033-080
Pyrrol-1-ylcarbamic acid tert-butyl ester 97%
Pyrrol-1-ylcarbamic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD09863891
CAS Number: 937046-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR