Ethyl 6-(trifluoromethyl)pyridazine-3-carboxylate 95%

Supplier: Ambeed
Warning

A345526-100MG A345526-250MG A345526-1G
77033-074EA 45 USD
77033-074 77594-030 77601-536
Ethyl 6-(trifluoromethyl)pyridazine-3-carboxylate 95%
Ethyl 6-(trifluoromethyl)pyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₇F₃N₂O₂
MDL Number: MFCD18072597
CAS Number: 1192155-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR