Methyl (4-cyanophenoxy)acetate 98%

Supplier: Ambeed
Warning

A345384-1G A345384-250MG A345384-5G
77015-634EA 6 USD
77015-634 77015-636 77015-638
Methyl (4-cyanophenoxy)acetate 98%
Methyl (4-cyanophenoxy)acetate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃
MDL Number: MFCD05855148
CAS Number: 272792-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR