Methyl 6-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A345296-250MG
A345296-50MG
77033-070EA
374.5
USD
77033-070
77033-072
Methyl 6-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate 95%
Methyl 6-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₀N₂O₂ MW: 190.19 g/mol |
MDL Number:
MFCD14706594 CAS Number: 872355-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...