2'-C-Methyluridine 97%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,3R,4R,5R)-3,4-Dihydroxy-5-(hydroxymethyl)-3-methyltetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-(2-C-Methyl-beta-D-ribofuranosyl)uracil

A344909-10G A344909-1G A344909-25G A344909-5G
76989-504EA 171.87 USD
76989-504 76989-506 76989-508 76989-510
2'-C-Methyluridine 97%
2'-C-Methyluridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O₆
MW: 258.23 g/mol
MDL Number: MFCD02682960
CAS Number: 31448-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR