tert-Butyl 5-bromo-3-methyl-1H-indazole-1-carboxylate 95%

Supplier: Ambeed
Danger

A344479-100MG A344479-1G A344479-250MG A344479-5G
77033-056EA 33 USD
77033-056 77033-058 77033-060 77033-062
tert-Butyl 5-bromo-3-methyl-1H-indazole-1-carboxylate 95%
tert-Butyl 5-bromo-3-methyl-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₅BrN₂O₂
MDL Number: MFCD24471372
CAS Number: 552331-49-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR