(S)-Piperidine-2-carboxylic acid hydrochloride 95%

Supplier: Ambeed
Warning

A344377-10G A344377-1G A344377-5G
77003-184EA 43.5 USD
77003-184 77003-186 77003-188
(S)-Piperidine-2-carboxylic acid hydrochloride 95%
(S)-Piperidine-2-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.6200 g/mol
MDL Number: MFCD00066139
CAS Number: 2133-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR