2-(2,4-Dimethylphenylsulfonamido)acetic acid 95%

Supplier: Ambeed
Warning

A343234-1G A343234-250MG A343234-5G
77015-622EA 57.01 USD
77015-622 77015-624 77015-626
2-(2,4-Dimethylphenylsulfonamido)acetic acid 95%
2-(2,4-Dimethylphenylsulfonamido)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD05986314
CAS Number: 670255-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR