2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate 97%

Supplier: Ambeed
Danger

Synonyms: TNTU, 2-(1,3-Dioxo-3a,4,7,7a-tetrahydro-1H-4,7-methanoisoindol-2(3H)-yl)-1,1,3,3-tetramethyluronium tetrafluoroborate

A342760-10G A342760-1G A342760-250MG A342760-25G
77003-172EA 35.01 USD
77003-172 77003-174 77003-176 77003-178
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate 97%
2-(5-Norbornene-2,3-dicarboximido)-1,1,3,3-tetramethyluronium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₄H₂₀BF₄N₃O₃
MW: 365.14 g/mol
MDL Number: MFCD00167753
CAS Number: 125700-73-4
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR