4-Methylpiperidine-4-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A342678-10G
A342678-250MG
A342678-1G
77015-618EA
445.58
USD
77015-618
77015-620
77594-002
4-Methylpiperidine-4-carboxylic acid hydrochloride 97%
4-Methylpiperidine-4-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₁₄ClNO₂ |
MDL Number:
MFCD18207802 CAS Number: 162648-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...