4-Bromo-2-methylisoindolin-1-one 95%

Supplier: Ambeed
Warning

A342403-250MG A342403-1G
77033-044EA 41 USD
77033-044 77872-509
4-Bromo-2-methylisoindolin-1-one 95%
4-Bromo-2-methylisoindolin-1-one

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MDL Number: MFCD11878013
CAS Number: 435273-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR