(2S,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolane-2-carboxylate 95+%

Supplier: Ambeed
Warning

A342369-100G A342369-1G A342369-250MG A342369-25G A342369-5G
77003-162EA 179.3 USD
77003-162 77003-164 77003-166 77003-168 77003-170
(2S,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolane-2-carboxylate 95+%
(2S,5R)-(1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl 5-(4-amino-5-fluoro-2-oxopyrimidin-1(2H)-yl)-1,3-oxathiolane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆FN₃O₄S
MW: 399.48 g/mol
MDL Number: MFCD09753149
CAS Number: 147126-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR