2-Amino-6-fluoropyridine 98%

Supplier: Ambeed
Warning

A342236-100G A342236-10G A342236-1G A342236-25G A342236-5G
77015-608EA 160.2 USD
77015-608 77015-610 77015-612 77015-614 77015-616
2-Amino-6-fluoropyridine 98%
2-Amino-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₅FN₂
MW: 112.11 g/mol
MDL Number: MFCD04114200
CAS Number: 1597-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR