1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate 95%

Supplier: Ambeed
Warning

A342219-100MG A342219-250MG
76989-460EA 59.42 USD
76989-460 76989-462
1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate 95%
1-(((4-Nitrophenoxy)carbonyl)oxy)ethyl isobutyrate

Precaución: for Research Use Only

Formula: C₁₃H₁₅NO₇
MW: 297.26 g/mol
MDL Number: MFCD22571566
CAS Number: 194995-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR