Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-[trans-4-(Boc-amino)cyclohexyl]acetate, Ethyl 2-(trans-4-((tert-butoxycarbonyl)amino)cyclohexyl)acetate, 2-[trans-4-(Boc-amino)cyclohexyl]acetic acid Ethyl ester, 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetic acid Ethyl ester

A341885-1G A341885-250MG A341885-5G
77015-602EA 49 USD
77015-602 77015-604 77015-606
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate 98%
Ethyl 2-[trans-4-[(tert-Butoxycarbonyl)amino]cyclohexyl]acetate

Precaución: for Research Use Only

Formula: C₁₅H₂₇NO₄
MDL Number: MFCD20491405
CAS Number: 946598-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR