1-Chloro-9,10-bis(phenylethynyl)anthracene 98%

Supplier: Ambeed
Warning

A341823-100G A341823-1G A341823-25G A341823-5G
77003-138EA 332 USD
77003-138 77003-140 77003-142 77003-144
1-Chloro-9,10-bis(phenylethynyl)anthracene 98%
1-Chloro-9,10-bis(phenylethynyl)anthracene

Precaución: for Research Use Only

Formula: C₃₀H₁₇Cl
MDL Number: MFCD00012047
CAS Number: 41105-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR