3-Bromo-1H-pyrazol-4-amine 97%

Supplier: Ambeed
Warning

A341572-100MG A341572-250MG A341572-1G
77015-592EA 55.17 USD
77015-592 77015-594 77550-986
3-Bromo-1H-pyrazol-4-amine 97%
3-Bromo-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₃H₄BrN₃
MW: 161.988 g/mol
MDL Number: MFCD26518979
CAS Number: 1374394-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR