4-Bromo-6-chloropicolinic acid 97%

Supplier: Ambeed
Warning

A341177-100MG A341177-1G A341177-250MG A341177-5G
77015-584EA 18 USD
77015-584 77015-586 77015-588 77015-590
4-Bromo-6-chloropicolinic acid 97%
4-Bromo-6-chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MDL Number: MFCD13189210
CAS Number: 1060805-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR