Dimethyl 3-bromopyridine-2,6-dicarboxylate 97%

Supplier: Ambeed
Warning

A340907-1G A340907-250MG
77015-570EA 61.54 USD
77015-570 77015-572 77015-574
Dimethyl 3-bromopyridine-2,6-dicarboxylate 97%
Dimethyl 3-bromopyridine-2,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₄
MDL Number: MFCD18258770
CAS Number: 316808-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR