1-Fluoro-2-methoxy-5-methyl-3-nitrobenzene 95%

Supplier: Ambeed
Warning

A340863-1G A340863-250MG
77015-560EA 185.66 USD
77015-560 77015-562
1-Fluoro-2-methoxy-5-methyl-3-nitrobenzene 95%
1-Fluoro-2-methoxy-5-methyl-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MW: 185.15 g/mol
MDL Number: MFCD22053532
CAS Number: 708-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR