3-Bromo-2,6-difluoropyridine 98%

Supplier: Ambeed
Warning

A340486-100MG A340486-10G A340486-1G A340486-250MG A340486-5G A340486-25G
76989-432EA 11 USD
76989-432 76989-434 76989-436 76989-438 76989-440 77612-952
3-Bromo-2,6-difluoropyridine 98%
3-Bromo-2,6-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrF₂N
MDL Number: MFCD11044274
CAS Number: 80392-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR