N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-D-Asp-OH, N-Fmoc-D-aspartic Acid
A340299-10G
A340299-1G
A340299-250MG
A340299-100G
A340299-5G
77033-030EA
62.6
USD
77033-030
77033-032
77033-034
77672-736
77672-738
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid
Precaución: for Research Use Only
Formula:
C₁₉H₁₇NO₆ MW: 355.35 g/mol |
MDL Number:
MFCD01318740 CAS Number: 136083-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...