N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine 98%

Supplier: Ambeed
Warning

Synonyms: N1,N1'-(1,4-Phenylene)bis(N4,N4-dibutyl-N1-(4-(dibutylamino)phenyl)benzene-1,4-diamine), N1,N1'-(1,4-Phenylene)bis[N4,N4-dibutyl-N1-[4-(dibutylamino)phenyl]benzene-1,4-diamine]

A340088-1G A340088-250MG
76989-428EA 22 USD
76989-428 76989-430
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine 98%
N,N,N',N'-Tetrakis[4-(dibutylamino)phenyl]-1,4-phenylenediamine

Precaución: for Research Use Only

Formula: C₆₂H₉₂N₆
MW: 921.46 g/mol
MDL Number: MFCD09832942
CAS Number: 4182-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR