2-(Methylamino)propan-1-ol 98%

Supplier: Ambeed
Danger

A339823-100MG A339823-250MG
77003-116EA 92.3 USD
77003-116 77003-118
2-(Methylamino)propan-1-ol 98%
2-(Methylamino)propan-1-ol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MDL Number: MFCD10686605
CAS Number: 27646-78-2
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR