Dimedone 98%

Supplier: Ambeed
Warning

Synonyms: 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethylcyclohexane-1,3-dione

A339754-1000G A339754-100G A339754-25G A339754-500G
77003-106EA 84 USD
77003-106 77003-108 77003-110 77003-112 77003-114
Dimedone 98%
Dimedone

Precaución: for Research Use Only

Formula: C₈H₁₂O₂
MDL Number: MFCD00001588
CAS Number: 126-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR