6-Chloro-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile 95%

Supplier: Ambeed
Danger

A339696-100MG A339696-1G A339696-250MG
77015-550EA 112.46 USD
77015-550 77015-552 77015-554
6-Chloro-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile 95%
6-Chloro-1H-pyrrolo[2,3-b]pyridine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MW: 177.59 g/mol
MDL Number: MFCD09880156
CAS Number: 1000340-76-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR