5-(Benzyloxy)-1H-indole-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A339640-1G A339640-250MG A339640-5G
77033-024EA 50.93 USD
77033-024 77033-026 77033-028
5-(Benzyloxy)-1H-indole-2-carboxylic acid 97%
5-(Benzyloxy)-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃NO₃
MDL Number: MFCD00047165
CAS Number: 6640-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR