4,5-Difluoro-2-nitrotoluene 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Difluoro-4-methyl-5-nitrobenzene

A339609-10G A339609-1G A339609-25G A339609-5G
77033-016EA 19 USD
77033-016 77033-018 77033-020 77033-022
4,5-Difluoro-2-nitrotoluene 98%
4,5-Difluoro-2-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD04973772
CAS Number: 127371-50-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR