4-Bromo-5-methyl-2-nitroanisole 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-methoxy-2-methyl-5-nitrobenzene

A339519-100MG A339519-10G A339519-1G A339519-250MG A339519-5G
76989-416EA 11.67 USD
76989-416 76989-418 76989-420 76989-422 76989-424
4-Bromo-5-methyl-2-nitroanisole 95%
4-Bromo-5-methyl-2-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.058 g/mol
MDL Number: MFCD23703052
CAS Number: 89978-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR