2-Cyano-3-fluorophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 2-Cyano-3-fluorobenzeneboronic acid pinacol ester

A339500-1G A339500-250MG A339500-5G
76989-410EA 26 USD
76989-410 76989-412 76989-414
2-Cyano-3-fluorophenylboronic acid pinacol ester 98%
2-Cyano-3-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MDL Number: MFCD09878541
CAS Number: 765916-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR