1-(4-((3'-Chloro-6-methoxy-[1,1'-biphenyl]-3-yl)methyl)phenyl)urea 98%

Supplier: Ambeed
Warning

A339395-10MG A339395-1MG A339395-50MG A339395-5MG A339395-100MG
76989-402EA 36 USD
76989-402 76989-404 76989-406 76989-408 77890-677
1-(4-((3'-Chloro-6-methoxy-[1,1'-biphenyl]-3-yl)methyl)phenyl)urea 98%
1-(4-((3'-Chloro-6-methoxy-[1,1'-biphenyl]-3-yl)methyl)phenyl)urea

Precaución: for Research Use Only

Formula: C₂₁H₁₉ClN₂O₂
MDL Number: MFCD22665725
CAS Number: 1155877-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR