tert-Butyl 2,4-dioxopyrrolidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A339257-1G A339257-250MG A339257-5G
76989-396EA 153.84 USD
76989-396 76989-398 76989-400
tert-Butyl 2,4-dioxopyrrolidine-1-carboxylate 95%
tert-Butyl 2,4-dioxopyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₃NO₄
MW: 199.20 g/mol
MDL Number: MFCD18914399
CAS Number: 182352-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR