tert-Butyl 3-iodo-6-methoxy-1H-indole-1-carboxylate 95%

Supplier: Ambeed
Warning

A339240-100MG A339240-1G A339240-250MG A339240-5G
77015-542EA 27.59 USD
77015-542 77015-544 77015-546 77015-548
tert-Butyl 3-iodo-6-methoxy-1H-indole-1-carboxylate 95%
tert-Butyl 3-iodo-6-methoxy-1H-indole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₆INO₃
MW: 373.18 g/mol
MDL Number: MFCD27923185
CAS Number: 1268167-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR