1-(2-Fluoro-3-methoxy-6-nitrophenyl)propan-2-one 97%

Supplier: Ambeed
Warning

A339225-100MG A339225-1G A339225-250MG
77015-536EA 25.47 USD
77015-536 77015-538 77015-540
1-(2-Fluoro-3-methoxy-6-nitrophenyl)propan-2-one 97%
1-(2-Fluoro-3-methoxy-6-nitrophenyl)propan-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀FNO₄
MW: 227.19 g/mol
MDL Number: MFCD11977234
CAS Number: 288385-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR