3-Hydroxy-5-methylpicolinic acid 98%

Supplier: Ambeed
Warning

A338999-1G A338999-250MG
77015-520EA 582.44 USD
77015-520 77015-522
3-Hydroxy-5-methylpicolinic acid 98%
3-Hydroxy-5-methylpicolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD13174803
CAS Number: 672957-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR