5H-Dibenzo[b,f]azepin-10(11H)-one 98%

Supplier: Ambeed
Warning

A338333-10G A338333-1G A338333-250MG A338333-5G
76989-388EA 730.97 USD
76989-388 76989-390 76989-392 76989-394
5H-Dibenzo[b,f]azepin-10(11H)-one 98%
5H-Dibenzo[b,f]azepin-10(11H)-one

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MW: 209.24 g/mol
MDL Number: MFCD00101078
CAS Number: 21737-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR