1-(2-Dimethylaminoethyl)-5-mercaptotetrazole 98%

Supplier: Ambeed
Danger

Synonyms: 1-(2-Dimethylaminoethyl)-1H-tetrazole-5-thiol, 1-(2-Dimethylaminoethyl)-5-mercapto-1,2,3,4-tetrazole

A338226-100G A338226-25G A338226-500G A338226-5G
76989-380EA 95.49 USD
76989-380 76989-382 76989-384 76989-386
1-(2-Dimethylaminoethyl)-5-mercaptotetrazole 98%
1-(2-Dimethylaminoethyl)-5-mercaptotetrazole

Precaución: for Research Use Only

Formula: C₅H₁₁N₅S
MW: 173.24 g/mol
MDL Number: MFCD00082923
CAS Number: 61607-68-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR