5-(2-Bromophenyl)isoxazole 97%

Supplier: Ambeed
Warning

A338152-1G A338152-5G
77003-098EA 63 USD
77003-098 77003-100
5-(2-Bromophenyl)isoxazole 97%
5-(2-Bromophenyl)isoxazole

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD02183530
CAS Number: 387358-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR