4,4'-Bis(5-methyl-2-benzoxazolyl)stilbene 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-(5-methylbenzo[d]oxazol-2-yl)phenyl)ethene

A337992-100G A337992-10G
77003-094EA 106.09 USD
77003-094 77003-096
4,4'-Bis(5-methyl-2-benzoxazolyl)stilbene 97%
4,4'-Bis(5-methyl-2-benzoxazolyl)stilbene

Precaución: for Research Use Only

Formula: C₃₀H₂₂N₂O₂
MW: 442.52 g/mol
MDL Number: MFCD00191316
CAS Number: 2397-00-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR